Learn More
Hach Company 1720D Drain Valve Assembly
Supplier:  Hach Company 5267500
Encompass
The 1720D Drain Valve Kit provides an easily-accessible means of draining the 1720D Series Turbidimeter for cleaning or other maintenance Opening the drain valve periodically to drain the turbidimeter body will reduce sediment collection and increase accuracy of readings
Catalog No. 50-002-07104