Learn More
Sino Biological Recombinant Human LILRB5/CD85c/LIR-8 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 17221H08H100
Encompass_Preferred
A DNA sequence encoding the human LILRB5 (NP_006831.1) (Met1-Gly458) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0003