Learn More
Sino Biological Recombinant Human TSHR Protein (ECD, His Tag)
Supplier:  Sino Biological 17299H08B50
Encompass_Preferred
A DNA sequence encoding the human TSHR (NP_000360.2) (Gly21-Gly413) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0007