Learn More
MSC Value Collection Oval Tube Axial Fan, 240 CFM, 1 ea
Catalog No. 5000506292
Encompass
This oval tube axial fan operates on 115V AC power, delivering an airflow of 240 CFM. Designed for efficient ventilation, it features a compact design suitable for various applications.
  • AC-powered operation
  • Current draw of 0.46 amp
  • Airflow of 240 CFM
  • Overall dimensions: 151 mm high, 172 mm wide, 51 mm deep
  • Includes fan, fan guard, and thermostatically controlled fan cord
Catalog No. 50-005-06292 Supplier MSC Supplier No. 44905800
This oval tube axial fan operates on 115V AC power, delivering an airflow of 240 CFM. Designed for efficient ventilation, it features a compact design suitable for various applications.
  • AC-powered operation
  • Current draw of 0.46 amp
  • Airflow of 240 CFM
  • Overall dimensions: 151 mm high, 172 mm wide, 51 mm deep
  • Includes fan, fan guard, and thermostatically controlled fan cord