Learn More
eMolecules​ 175278-59-8 | 4,6-Dimethyl-1,3,5-triazin-2-amine hydrate | AA Blocks LLC | MFCD00663061 | 142.162 | C5H10N4O | 0.000 | O.Cc1nc(C)nc(N)n1 | 1g | 913032006
Supplier:  eMolecules​ 913032006
Encompass_Preferred
4,6-Dimethyl-1,3,5-triazin-2-amine hydrate | AA Blocks LLC | 175278-59-8 | MFCD00663061 | 142.162 | C5H10N4O | 0.000 | O.Cc1nc(C)nc(N)n1 | 1g | 913032006
Catalog No. NC3966031