Learn More
Alkali Scientific 175cm² Cell Culture Flasks, Vent Cap, Non-Treated, Sterile, 5/pk, 40/cs
SDP
Catalog No. 502389214
Encompass_Preferred
These Cell Culture Flasks are non-treated, vented and sterile. They come packaged in zip bags and are high clarity, 100% virgin polystyrene. They are stackable, sterilized by E-Beam, non-pyrogenic, and DNase/RNase free. They have frosted writing and clear graduations and clear lot number for batch traceability.
Catalog No. 50-238-9214 Supplier Alkali Scientific Supplier No. TL33
These Cell Culture Flasks are non-treated, vented and sterile. They come packaged in zip bags and are high clarity, 100% virgin polystyrene. They are stackable, sterilized by E-Beam, non-pyrogenic, and DNase/RNase free. They have frosted writing and clear graduations and clear lot number for batch traceability.