Learn More
Chemglass Life Sciences 175cm² Cell Culture Flask, Plug Seal Cap, Non-Treated, Sterile, 5/pk, 40/cs
SDP
Catalog No. 501897665
Encompass_Preferred
Made from high clarity, 100% virgin polystyreneSterilized by E-beamClear lot number for batch traceabilityIndividually plastic wrappedNon-Pyrogenic, DNase/RNase-FreeAngled neck for easy accessFrosted writing and clear graduationsNotched bottom for slip-free stackingSolid Plug Seal Caps
Catalog No. 50-189-7665 Supplier Chemglass Life Sciences Supplier No. CGN2408P175
Made from high clarity, 100% virgin polystyreneSterilized by E-beamClear lot number for batch traceabilityIndividually plastic wrappedNon-Pyrogenic, DNase/RNase-FreeAngled neck for easy accessFrosted writing and clear graduationsNotched bottom for slip-free stackingSolid Plug Seal Caps