Learn More
Chemglass Life Sciences 175cm² Cell Culture Flask, TC, Vent Cap, Sterile, 5/pk, 40/cs
SDP
Catalog No. 501897657
Encompass_Preferred
Made from High clarity, 100% virgin polystyreneSterilized by E-beamVacuum plasma tissue culture treatmentClear lot number for batch traceabilityIndividually plastic wrappedNon-Pyrogenic, DNase/RNase FreeAngled neck for easy accessFrosted writing and clear graduationsNotched bottom for slip-free stackingVented caps (with breathable membrane)Cell culture flasks are available in cell growth areas ranging from 25cm¿ to 225cm¿. Tissue culture treated with a vent cap. Vent caps with 0.22 ?m hydrophobic filters for gas exchange.
Catalog No. 50-189-7657 Supplier Chemglass Life Sciences Supplier No. CGN2406175
Made from High clarity, 100% virgin polystyreneSterilized by E-beamVacuum plasma tissue culture treatmentClear lot number for batch traceabilityIndividually plastic wrappedNon-Pyrogenic, DNase/RNase FreeAngled neck for easy accessFrosted writing and clear graduationsNotched bottom for slip-free stackingVented caps (with breathable membrane)Cell culture flasks are available in cell growth areas ranging from 25cm¿ to 225cm¿. Tissue culture treated with a vent cap. Vent caps with 0.22 ?m hydrophobic filters for gas exchange.