Learn More
NEST SCIENTIFIC INC 175CM2 CELL CULTURE FLK
Catalog No. NC3429290
Encompass_Preferred
Nest Scientific tissue culture flasks are molded from crystal clear USP grade VI virgin polystyrene. They are plasma etched (TC treated) to support adherent cell lines. Feature a vented cap with a USP grade VI 0.22uM PTFE membrane for gas exchange. Flask upper and lower are ultrasonically welded together with no adhesives that may leach. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Extensively tested for leachables/extractables per ISO 10993. This flask is among our best selling items worldwide. ~20% of US brands on US market private label this flask from Nest. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.
Catalog No. NC3429290 Supplier NEST SCIENTIFIC INC Supplier No. 709003PK
Nest Scientific tissue culture flasks are molded from crystal clear USP grade VI virgin polystyrene. They are plasma etched (TC treated) to support adherent cell lines. Feature a vented cap with a USP grade VI 0.22uM PTFE membrane for gas exchange. Flask upper and lower are ultrasonically welded together with no adhesives that may leach. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Extensively tested for leachables/extractables per ISO 10993. This flask is among our best selling items worldwide. ~20% of US brands on US market private label this flask from Nest. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.