Learn More
Ace Glass, Inc. Solvent reservoir, 175mL, #27 stopper joint top, 19/22 bottom, 4mm PTFE stopcock (8224-12)
SDP
Catalog No. 50972410
Encompass_Preferred
175mL capacity neutral oil determination chromatography reservoir has a 4mm PTFE stopcock, a 19/22 standard taper male joint at the bottom, and a #27 stopper joint at the top. Meets A.O.C.S. CA9F-57 specifications.
Catalog No. 50-972-410 Supplier Ace Glass, Inc. Supplier No. 586902
175mL capacity neutral oil determination chromatography reservoir has a 4mm PTFE stopcock, a 19/22 standard taper male joint at the bottom, and a #27 stopper joint at the top. Meets A.O.C.S. CA9F-57 specifications.