Learn More
Fluidigm Corp 176YB X8 ANTIBODY LABELING
SDP
Catalog No. NC0521840
Encompass
This Maxpar kit lets you conjugate IgG antibodies with the 176Yb metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.
Catalog No. NC0521840 Supplier Fluidigm Corp Supplier No. 201176A
This Maxpar kit lets you conjugate IgG antibodies with the 176Yb metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.