Learn More
Cayman Chemical 17a 20-DIhydroxy-4-pregn-3 1mg
SDP
Catalog No. 502970251
Encompass_Preferred
An endogenous, maturation-inducing steroid that at 1 UG/ml induces germinal vesicle breakdown in oocytes from several teleost species; can also influence spermiation by stimulating milt production when administered at 5 mg/kg to various male teleosts
Catalog No. 50-297-0251 Supplier Cayman Chemical Supplier No. 161461 MG
An endogenous, maturation-inducing steroid that at 1 UG/ml induces germinal vesicle breakdown in oocytes from several teleost species; can also influence spermiation by stimulating milt production when administered at 5 mg/kg to various male teleosts