Learn More
Cayman Chemical 17α,20β-Dihydroxy-4-pregnen-3-one, Purity: ≥95%, CAS: 1662-06-2, 10 mg
SDP
Catalog No. 502971572
Encompass_Preferred
An endogenous, maturation-inducing steroid that at 1 µg/ml induces germinal vesicle breakdown in oocytes from several teleost species; can also influence spermiation by stimulating milt production when administered at 5 mg/kg to various male teleosts
Catalog No. 50-297-1572 Supplier Cayman Chemical Supplier No. 1614610 MG
An endogenous, maturation-inducing steroid that at 1 µg/ml induces germinal vesicle breakdown in oocytes from several teleost species; can also influence spermiation by stimulating milt production when administered at 5 mg/kg to various male teleosts