Learn More
Cayman Chemical 17a-hydroxy Pregnolon 1g
SDP
Catalog No. 502941940
Encompass_Preferred
A metabolite of pregnenolone; a precursor in the biosynthesis of DHEA; plasma levels are elevated in patients with type II 3β-hydroxysteroid dehydrogenase deficiency
Catalog No. 50-294-1940 Supplier Cayman Chemical Supplier No. 291811 G
A metabolite of pregnenolone; a precursor in the biosynthesis of DHEA; plasma levels are elevated in patients with type II 3β-hydroxysteroid dehydrogenase deficiency