Learn More
Cayman Chemical 17a-hydroxy Pregnolon 250mg
SDP
Catalog No. 502941358
Encompass_Preferred
A metabolite of pregnenolone; a precursor in the biosynthesis of DHEA; plasma levels are elevated in patients with type II 3β-hydroxysteroid dehydrogenase deficiency
Catalog No. 50-294-1358 Supplier Cayman Chemical Supplier No. 29181250 MG
A metabolite of pregnenolone; a precursor in the biosynthesis of DHEA; plasma levels are elevated in patients with type II 3β-hydroxysteroid dehydrogenase deficiency