Learn More
Cayman Chemical 17a-hydroxy Pregnolon 500mg
SDP
Catalog No. 502939973
Encompass_Preferred
A metabolite of pregnenolone; a precursor in the biosynthesis of DHEA; plasma levels are elevated in patients with type II 3β-hydroxysteroid dehydrogenase deficiency
Catalog No. 50-293-9973 Supplier Cayman Chemical Supplier No. 29181500 MG
A metabolite of pregnenolone; a precursor in the biosynthesis of DHEA; plasma levels are elevated in patients with type II 3β-hydroxysteroid dehydrogenase deficiency