Learn More
Medchemexpress LLC 17a-Hydroxypregnenolone (17-OHP5) | 387-79-1 | 99.61% | 332.48 | 100 MG
SDP
Catalog No. 5000437940
Encompass_Preferred
17a-Hydroxypregnenolone (17-OHP5) is a metabolite of Pregnenolone and a precursor in the biosynthesis of Dehydroepiandrosterone. It acts as a substrate for the enzyme 3β-hydroxysteroid dehydrogenase (3β-HSD). This compound is formed from pregnenolone by cytochrome P450 (CYP) isoform CYP17A1 hydroxylase activity and subsequently converted to dehydroepiandrosterone by CYP17A1-mediated 17,20-lyase activity. Levels of 17a-hydroxypregnenolone are elevated in patients with type II 3β-hydroxysteroid dehydrogenase deficiency, which is a form of congenital adrenal hyperplasia.
  • Metabolite of pregnenolone
  • Precursor in dehydroepiandrosterone biosynthesis
  • Substrate for 3β-hydroxysteroid dehydrogenase
  • Formed by CYP17A1 hydroxylase activity
  • Converted to dehydroepiandrosterone by CYP17A1-mediated 17,20-lyase activity
  • Elevated in type II 3β-hydroxysteroid dehydrogenase deficiency
Catalog No. 50-004-37940 Supplier Medchemexpress LLC Supplier No. HY113263100MG
17a-Hydroxypregnenolone (17-OHP5) is a metabolite of Pregnenolone and a precursor in the biosynthesis of Dehydroepiandrosterone. It acts as a substrate for the enzyme 3β-hydroxysteroid dehydrogenase (3β-HSD). This compound is formed from pregnenolone by cytochrome P450 (CYP) isoform CYP17A1 hydroxylase activity and subsequently converted to dehydroepiandrosterone by CYP17A1-mediated 17,20-lyase activity. Levels of 17a-hydroxypregnenolone are elevated in patients with type II 3β-hydroxysteroid dehydrogenase deficiency, which is a form of congenital adrenal hyperplasia.
  • Metabolite of pregnenolone
  • Precursor in dehydroepiandrosterone biosynthesis
  • Substrate for 3β-hydroxysteroid dehydrogenase
  • Formed by CYP17A1 hydroxylase activity
  • Converted to dehydroepiandrosterone by CYP17A1-mediated 17,20-lyase activity
  • Elevated in type II 3β-hydroxysteroid dehydrogenase deficiency