Learn More
Medchemexpress LLC 17a-Hydroxypregnenolone-d3 | 105078-92-0 | 335.50 | 1 MG
SDP
Catalog No. 5000438554
Encompass_Preferred
17a-Hydroxypregnenolone-d3 (17-OHP5-d3) is a deuterium labeled metabolite of Pregnenolone and a precursor in the biosynthesis of Dehydroepiandrosterone. It acts as a substrate for the enzyme 3β-hydroxysteroid dehydrogenase (3β-HSD).
  • Deuterium labeled 17a-hydroxypregnenolone.
  • Metabolite of pregnenolone.
  • Precursor in the biosynthesis of dehydroepiandrosterone.
  • Substrate for the enzyme 3β-hydroxysteroid dehydrogenase (3β-HSD).
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-38554 Supplier Medchemexpress LLC Supplier No. HY113263S1MG
17a-Hydroxypregnenolone-d3 (17-OHP5-d3) is a deuterium labeled metabolite of Pregnenolone and a precursor in the biosynthesis of Dehydroepiandrosterone. It acts as a substrate for the enzyme 3β-hydroxysteroid dehydrogenase (3β-HSD).
  • Deuterium labeled 17a-hydroxypregnenolone.
  • Metabolite of pregnenolone.
  • Precursor in the biosynthesis of dehydroepiandrosterone.
  • Substrate for the enzyme 3β-hydroxysteroid dehydrogenase (3β-HSD).
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.