Learn More
Medchemexpress LLC 17a-Hydroxypregnenolone-d3 | 105078-92-0 | 335.50 | 10 MG
SDP
Catalog No. 5000437869
Encompass_Preferred
17a-Hydroxypregnenolone-d3 is a deuterium-labeled compound that functions as a metabolite of Pregnenolone and a precursor in the biosynthesis of Dehydroepiandrosterone. It also serves as a substrate for the enzyme 3β-hydroxysteroid dehydrogenase. The incorporation of stable heavy isotopes like deuterium allows its use as a tracer in drug development to influence pharmacokinetic and metabolic profiles.
  • Deuterium labeled compound
  • Metabolite of Pregnenolone
  • Precursor in biosynthesis of Dehydroepiandrosterone
  • Substrate for enzyme 3β-hydroxysteroid dehydrogenase
  • Used as a tracer in drug development processes
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-37869 Supplier Medchemexpress LLC Supplier No. HY113263S10MG
17a-Hydroxypregnenolone-d3 is a deuterium-labeled compound that functions as a metabolite of Pregnenolone and a precursor in the biosynthesis of Dehydroepiandrosterone. It also serves as a substrate for the enzyme 3β-hydroxysteroid dehydrogenase. The incorporation of stable heavy isotopes like deuterium allows its use as a tracer in drug development to influence pharmacokinetic and metabolic profiles.
  • Deuterium labeled compound
  • Metabolite of Pregnenolone
  • Precursor in biosynthesis of Dehydroepiandrosterone
  • Substrate for enzyme 3β-hydroxysteroid dehydrogenase
  • Used as a tracer in drug development processes
  • Potential to affect pharmacokinetic and metabolic profiles of drugs