Learn More
Medchemexpress LLC 17a-hydroxypregnenolone-d3 | 105078-92-0 | MFCD01317456 | 99.0% | 335.50 g/mol | C21H29D3O3 | 5 MG
SDP
Catalog No. 5000264716
Encompass_Preferred
17a-Hydroxypregnenolone-d3 is the deuterium-labeled (d3) isotopologue of 17α-hydroxypregnenolone, intended as a stable-isotope internal standard and reference material for quantitative steroid analysis and metabolic studies. It is supplied as a high-purity solid (99.0%) with molecular formula C21H29D3O3 and molecular weight 335.50 g/mol.
  • Labeled internal standard for quantitative LC-MS/MS and GC-MS assays.
  • High chemical and isotopic purity for reliable quantitation.
  • Provided as a dry solid for controlled dissolution and storage.
  • Comes with COA and SDS to support analytical validation.
  • Available in small mg-scale packs suitable for laboratory use.
Catalog No. 50-002-64716 Supplier Medchemexpress LLC Supplier No. HY113263S5MG
17a-Hydroxypregnenolone-d3 is the deuterium-labeled (d3) isotopologue of 17α-hydroxypregnenolone, intended as a stable-isotope internal standard and reference material for quantitative steroid analysis and metabolic studies. It is supplied as a high-purity solid (99.0%) with molecular formula C21H29D3O3 and molecular weight 335.50 g/mol.
  • Labeled internal standard for quantitative LC-MS/MS and GC-MS assays.
  • High chemical and isotopic purity for reliable quantitation.
  • Provided as a dry solid for controlled dissolution and storage.
  • Comes with COA and SDS to support analytical validation.
  • Available in small mg-scale packs suitable for laboratory use.