Learn More
Cayman Chemical 17a-Hydroxyprogesteron 10g
SDP
Catalog No. 502950196
Encompass_Preferred
A steroid and precursor in the synthesis of adrenal androgens; binds to the glucocorticoid receptor in a cell-free assay (IC50 = 82 nM for the human receptor) and induces glucocorticoid receptor transactivation in HEK293 cells (EC50 = 1.2 µM); inhibits aldosterone-induced transactivation of the mineralocorticoid receptor in cell-based assays from 0.005-1 µM; serum levels are increased in patients with congenital adrenal hyperplasia
Catalog No. 50-295-0196 Supplier Cayman Chemical Supplier No. 3315410 G
A steroid and precursor in the synthesis of adrenal androgens; binds to the glucocorticoid receptor in a cell-free assay (IC50 = 82 nM for the human receptor) and induces glucocorticoid receptor transactivation in HEK293 cells (EC50 = 1.2 µM); inhibits aldosterone-induced transactivation of the mineralocorticoid receptor in cell-based assays from 0.005-1 µM; serum levels are increased in patients with congenital adrenal hyperplasia