Learn More
Innovative Reseach Of America 17β-Estradiol Pellet, 90-Day Release, 0.1 mg
SDP
Catalog No. NC3709736
Encompass
The Innovative Research of America NE-121-0.1MG is a 90-day release 17β-Estradiol pellet designed for sustained hormone delivery in laboratory animal research. Utilizing a Matrix-Driven Delivery system, it offers continuous release of 0.1 mg 17β-Estradiol, providing a standardized drug administration method that enhances research reproducibility by reducing the need for frequent injections.
Catalog No. NC3709736 Supplier Innovative Reseach Of America Supplier No. NE1210.1MG
The Innovative Research of America NE-121-0.1MG is a 90-day release 17β-Estradiol pellet designed for sustained hormone delivery in laboratory animal research. Utilizing a Matrix-Driven Delivery system, it offers continuous release of 0.1 mg 17β-Estradiol, providing a standardized drug administration method that enhances research reproducibility by reducing the need for frequent injections.