Learn More
Innovative Reseach Of America 17β-Estradiol 0.72 mg Pellets, Sustained Release 60 Days, Subcutaneous Implantation
SDP
Catalog No. NC9032951
Encompass
The Innovative Research of America 17β-Estradiol 0.72 mg Pellets provide a reliable method for sustained hormone delivery, designed for in vivo research. Utilizing the Matrix-Driven Delivery (MDD) system, these pellets ensure consistent hormone levels over 60 days, making them essential for studies in endocrinology, oncology, and reproductive biology.
Catalog No. NC9032951 Supplier Innovative Reseach Of America Supplier No. SE1210.72MG_60_25P
The Innovative Research of America 17β-Estradiol 0.72 mg Pellets provide a reliable method for sustained hormone delivery, designed for in vivo research. Utilizing the Matrix-Driven Delivery (MDD) system, these pellets ensure consistent hormone levels over 60 days, making them essential for studies in endocrinology, oncology, and reproductive biology.