Learn More
Innovative Reseach Of America 17β-Estradiol Pellet, Sustained Release, Biodegradable Matrix, 60 Days
SDP
Catalog No. NC0388908
Encompass
17β-Estradiol Slow-Release Pellet System delivers 0.36 mg of active ingredient consistently over 60 days. Designed to enhance experimental reliability in in vivo animal research while reducing the need for frequent injections. The biodegradable Matrix-Driven Delivery (MDD) system minimizes variability, making it ideal for studies in endocrinology, oncology, and reproductive biology. Pack contains 25 pellets.
Catalog No. NC0388908 Supplier Innovative Reseach Of America Supplier No. SE1210.36MG_60_25P
17β-Estradiol Slow-Release Pellet System delivers 0.36 mg of active ingredient consistently over 60 days. Designed to enhance experimental reliability in in vivo animal research while reducing the need for frequent injections. The biodegradable Matrix-Driven Delivery (MDD) system minimizes variability, making it ideal for studies in endocrinology, oncology, and reproductive biology. Pack contains 25 pellets.