Learn More
Innovative Reseach Of America 17ß-Estradiol 90 Day Release Pellet, Continuous Release, Biodegradable, 0.5 mg each
SDP
Catalog No. NC1970566
Encompass
The 17ß-Estradiol 90 Day Release Pellet by Innovative Research of America offers a stable, long-term delivery system for researchers studying hormone-dependent conditions. Its Matrix-Driven Delivery system ensures consistent hormone release, eliminating variability from injections. This biodegradable pellet lasts 90 days, making it ideal for animal studies in reproductive biology and tumor growth. This product prioritizes safe, reliable research practices while being economical and effective.
Catalog No. NC1970566 Supplier Innovative Reseach Of America Supplier No. NE1210.5MG9025P
The 17ß-Estradiol 90 Day Release Pellet by Innovative Research of America offers a stable, long-term delivery system for researchers studying hormone-dependent conditions. Its Matrix-Driven Delivery system ensures consistent hormone release, eliminating variability from injections. This biodegradable pellet lasts 90 days, making it ideal for animal studies in reproductive biology and tumor growth. This product prioritizes safe, reliable research practices while being economical and effective.