Learn More
Enzo Life Sciences 17beta-Estradiol high sensitivity ELISA kit (5x96wells)
SDP
Catalog No. 502006897
Encompass_Preferred
Highly sensitive ELISA, detecting as little as 14 pg/ml of 17β-Estradiol. The 17β-Estradiol (serum/plasma) EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Alternative name: Estrogen. Sensitivity: 14.0 pg/ml (range 15.6 - 1,000 pg/ml). Use/Stability: The Estradiol-17β Antibody and Conjugate are stable at -20°C. All other components of this kit are stable at +4°C until the kit's expiration date. The Antibody and Conjugate should be stored at -20°C upon receipt. Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .
Catalog No. 50-200-6897 Supplier Enzo Life Sciences Supplier No. ADI901174
Add to Cart
Add to Cart
Highly sensitive ELISA, detecting as little as 14 pg/ml of 17β-Estradiol. The 17β-Estradiol (serum/plasma) EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Alternative name: Estrogen. Sensitivity: 14.0 pg/ml (range 15.6 - 1,000 pg/ml). Use/Stability: The Estradiol-17β Antibody and Conjugate are stable at -20°C. All other components of this kit are stable at +4°C until the kit's expiration date. The Antibody and Conjugate should be stored at -20°C upon receipt. Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .