Learn More
Medchemexpress LLC 17beta-dihydroequilin-2,4,16,16-d4 | 350819-99-7 | MFCD01074213 | 99.0% | 272.37 g·mol-1 | C18H16D4O2 | 5 MG
SDP
Catalog No. 5000222981
Encompass_Preferred
β-Dihydroequilin-d4 is the deuterium-labeled isotopologue of β-dihydroequilin intended for use as an analytical standard and internal standard in mass spectrometry and quantitative research. Supplied as a solid high-purity isotopic reference (CAS 350819-99-7), it supports assay development, isotopic tracing, and accurate quantification in laboratory workflows.
  • High isotopic enrichment for accurate mass spectrometry quantification.
  • Purity reported at approximately 99.0% for analytical use.
  • Supplied as a solid for convenient handling and long-term storage.
  • Suitable as an internal standard in LC-MS/MS and GC-MS assays.
  • Supports isotopic tracing and quantitative method development.
Catalog No. 50-002-22981 Supplier Medchemexpress LLC Supplier No. HY138112S25MG
β-Dihydroequilin-d4 is the deuterium-labeled isotopologue of β-dihydroequilin intended for use as an analytical standard and internal standard in mass spectrometry and quantitative research. Supplied as a solid high-purity isotopic reference (CAS 350819-99-7), it supports assay development, isotopic tracing, and accurate quantification in laboratory workflows.
  • High isotopic enrichment for accurate mass spectrometry quantification.
  • Purity reported at approximately 99.0% for analytical use.
  • Supplied as a solid for convenient handling and long-term storage.
  • Suitable as an internal standard in LC-MS/MS and GC-MS assays.
  • Supports isotopic tracing and quantitative method development.