Learn More
Innovative Reseach Of America 17BETA ESTRADIOL 1.7MG 25PELL
SDP
Catalog No. NC9145499
Encompass
17beta-Estradiol, For tumor growth and regression in immuno-deficient mice, 90-day release pellet, Total Dose: 1.7mg/PK, Pellet Size: 3mm
Catalog No. NC9145499 Supplier Innovative Reseach Of America Supplier No. NE1211.7MG_90_25P
17beta-Estradiol, For tumor growth and regression in immuno-deficient mice, 90-day release pellet, Total Dose: 1.7mg/PK, Pellet Size: 3mm