Learn More
Enzo Life Sciences 17beta-Estradiol ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901008
Encompass_Preferred
Highly sensitive ELISA, measuring as little as 28.5 pg/ml of 17β-Estradiol in < 3 hours. The 17β-Estradiol EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. This kit offers specific and save non-radioactive quantification of estradiol with ready-to-use liquid color-coded reagents to reduce error. Alternative name: Estrogen. Sensitivity: 28.5 pg/ml (range 29.3 - 30,000 pg/ml). Assay Time: <3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent . Content: GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent.
Catalog No. 50-200-6883