Learn More
Medchemexpress LLC 17beta-methoxy-3-propoxyestra-1,3,5(10)-triene | 39219-28-8 | MFCD00867247 | 99.9% | 328.49 | C22H32O2 | 200 MG
SDP
Catalog No. 5000241996
Encompass_Preferred
Promestriene is a synthetic diethyl-ether derivative of estradiol that acts as a locally effective estrogen. It is used in research to model topical estrogenic activity and has reported efficacy against vaginal atrophy while showing minimal systemic absorption. Provided as a high-purity research reagent for in vitro and analytical applications; not for human use.
  • Locally active estrogenic agent.
  • Used to study vaginal atrophy and estrogen receptor activity.
  • Low systemic absorption when applied topically.
  • Chemical formula C22H32O2; molecular weight 328.49.
  • High purity (≈99.9%) as supplied.
  • For research use only; not for clinical use.
Catalog No. 50-002-41996 Supplier Medchemexpress LLC Supplier No. HY108293200MG
Promestriene is a synthetic diethyl-ether derivative of estradiol that acts as a locally effective estrogen. It is used in research to model topical estrogenic activity and has reported efficacy against vaginal atrophy while showing minimal systemic absorption. Provided as a high-purity research reagent for in vitro and analytical applications; not for human use.
  • Locally active estrogenic agent.
  • Used to study vaginal atrophy and estrogen receptor activity.
  • Low systemic absorption when applied topically.
  • Chemical formula C22H32O2; molecular weight 328.49.
  • High purity (≈99.9%) as supplied.
  • For research use only; not for clinical use.