Learn More
Perkin Elmer US LLC 17-Element Solution, 5% HNO3, 1,000 µg/mL, 500 mL
Catalog No. 502103705
Encompass_Preferred
Details Concentration Element 1,000 µg/mL As, Ba, Be, Cd, Cr, Co, Cu, La, Pb, Li, Mn, Ni, Sc, Sr, V, Y, Zn
Catalog No. 50-210-3705 Supplier Perkin Elmer US LLC Supplier No. N9307116
Add to Cart
Add to Cart
Details Concentration Element 1,000 µg/mL As, Ba, Be, Cd, Cr, Co, Cu, La, Pb, Li, Mn, Ni, Sc, Sr, V, Y, Zn