Learn More
SKYLINE CENTER Item #W47 Bulk 17" X 17" Cloth - 100ct.
Catalog No. NC9701418
Encompass
17" X 17" CLOTH Uses: Cleaning - guns, firearms, melt flow index, melt plastometers, capillary rheometers, optics, laboratory equipment.
This item is not returnable. View return policy
Catalog No. NC9701418 Supplier SKYLINE CENTER Supplier No. 47
17" X 17" CLOTH Uses: Cleaning - guns, firearms, melt flow index, melt plastometers, capillary rheometers, optics, laboratory equipment.