Learn More
Beckman Coulter 17ML STERILE+CERT FREE TUBE
Catalog No. NC2151367 Shop All Beckman Coulter Products
Encompass_Preferred
17mL sterile+Cert free open top, thin wall Ultra clear tube
This item is not returnable. View return policy
Catalog No. NC2151367 Supplier Beckman Coulter Supplier No. C14297
17mL sterile+Cert free open top, thin wall Ultra clear tube