Learn More
Ace Glass, Inc. 17MM GRADED SEALS 17MM GRADED SEALS
SDP
Catalog No. 50974599
Encompass
17mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-599 Supplier Ace Glass, Inc. Supplier No. 845528
17mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.