Learn More
MSC 3M Powered Air-Purifying Respirator Kit, Headcover, HEPA Filter, Polypropylene
Catalog No. 502835009
Encompass
This Optimair TL Kit provides comprehensive respiratory protection as a Powered Air-Purifying Respirator (PAPR) system. Designed for applications such as powder handling, it features a polypropylene headcover and a high-efficiency HEPA filter, ensuring safety and comfort in demanding environments.
  • Comprehensive Powered Air-Purifying Respirator (PAPR) system.
  • Designed for powder handling applications.
  • Features polypropylene headgear with adjustable suspension.
  • Equipped with a HEPA filter and breathing tube.
  • Includes a hood with integrated head suspension.
  • Kit contains battery, charger, and easy clean belt.
  • Comes with prefilters, spark arrestors, and an airflow indicator.
  • Includes filter cover, breathing tube cover, and battery attachment tool.
  • Supplied with a storage and cleaning kit.
Catalog No. 50-283-5009 Supplier MSC Supplier No. 46721353
This Optimair TL Kit provides comprehensive respiratory protection as a Powered Air-Purifying Respirator (PAPR) system. Designed for applications such as powder handling, it features a polypropylene headcover and a high-efficiency HEPA filter, ensuring safety and comfort in demanding environments.
  • Comprehensive Powered Air-Purifying Respirator (PAPR) system.
  • Designed for powder handling applications.
  • Features polypropylene headgear with adjustable suspension.
  • Equipped with a HEPA filter and breathing tube.
  • Includes a hood with integrated head suspension.
  • Kit contains battery, charger, and easy clean belt.
  • Comes with prefilters, spark arrestors, and an airflow indicator.
  • Includes filter cover, breathing tube cover, and battery attachment tool.
  • Supplied with a storage and cleaning kit.