Learn More
Cayman Chemical 17-phenyl trinor Prostaglandin F2α, Purity: ≥98%, CAS: 38344-08-0, 10 mg
SDP
Catalog No. 502967943
Encompass_Preferred
A metabolically stable analog of PGF2α and potent FP receptor agonist (Ki = 1.1 nM at the rat FP receptor)
Catalog No. 50-296-7943 Supplier Cayman Chemical Supplier No. 1681010 MG
A metabolically stable analog of PGF2α and potent FP receptor agonist (Ki = 1.1 nM at the rat FP receptor)