Learn More
Cayman Chemical 17phenl trInor ProstglndI 10mg
SDP
Supplier:  Cayman Chemical 1682310 MG
Encompass_Preferred
An F-series PG analog in which the C-1 carboxyl group has been modified to an N-diethyl amide; dialkyl amides such as a 17-p-PGF2α-NEt2 are inert to corneal amidase activity, and are not converted in any detectable amount to the corresponding free acids
Catalog No. 50-296-5302