Learn More
Cayman Chemical 17phenl trInor ProstglndI 10mg
SDP
Catalog No. 502965309
Encompass_Preferred
An isopropyl ester of the free acid PG which corresponds to bimatoprost; a potent ocular hypotensive agent, lowering the IOP 1.3 mm Hg below the level achieved by latanoprost, but with significantly more irritation
Catalog No. 50-296-5309 Supplier Cayman Chemical Supplier No. 1682410 MG
An isopropyl ester of the free acid PG which corresponds to bimatoprost; a potent ocular hypotensive agent, lowering the IOP 1.3 mm Hg below the level achieved by latanoprost, but with significantly more irritation