Learn More
Cayman Chemical 17-phenyl trinor Prostaglandin F2α ethyl amide, Purity: ≥97%, CAS: 155206-00-1, 50 mg
SDP
Catalog No. 502967942
Encompass_Preferred
An F-series prostaglandin and an FP receptor agonist; induces calcium mobilization in 3T3-L1 fibroblasts expressing the human receptors (EC50 = 2.2 µM); ocular administration reduces IOP in normotensive dogs and ocular hypertensive monkeys at 0.03% w/v
Catalog No. 50-296-7942 Supplier Cayman Chemical Supplier No. 1682050 MG
An F-series prostaglandin and an FP receptor agonist; induces calcium mobilization in 3T3-L1 fibroblasts expressing the human receptors (EC50 = 2.2 µM); ocular administration reduces IOP in normotensive dogs and ocular hypertensive monkeys at 0.03% w/v