Learn More
Cayman Chemical 17phenyl trInor Prostglnd 10mg
SDP
Supplier:  Cayman Chemical 1353210 MG
Encompass_Preferred
Derived from 17-phenyl trinor PGE2, a synthetic analog of PGE2 that acts as an agonist of EP1 and EP3 in mice (Ki = 14 and 3.7 nM, respectively) and EP1, EP3, and EP4 in rats (Ki = 25, 4.3, and 54 nM, respectively); the ethyl amide group increases lipid solubility, lowering the effective concentration
Catalog No. 50-295-5835