Learn More
Cayman Chemical 17phenyl trInor ProstglnI 10mg
SDP
Catalog No. 502954032
Encompass_Preferred
The 1,15 lactone of 17-phenyl-trinor PGF2α with potential for suitable pharmacologic activity and an improved side-effect profile over current glaucoma therapeutics
Catalog No. 50-295-4032 Supplier Cayman Chemical Supplier No. 1399210 MG
The 1,15 lactone of 17-phenyl-trinor PGF2α with potential for suitable pharmacologic activity and an improved side-effect profile over current glaucoma therapeutics