Learn More
Cayman Chemical 17-phenyl trinor Prostaglandin E2, Purity: ≥98%, CAS: 38315-43-4, 10 mg
SDP
Catalog No. 502971443
Encompass_Preferred
17-phenyl trinor PGE2 is a synthetic analog of PGE2. It is an EP1 and EP3 receptor agonist.{3318,1025} 17-phenyl trinor PGE2 is an EP1 and EP3 receptor agonist.1,2 It causes contraction of the guinea pig ileum at a concentration of 11 µM.{3318} It is slightly less potent than PGE2 in stimulating gerbil colon and rat uterus.{1025} With an ED50 value of 350 µg/kg, 17-phenyl trinor PGE2 is 4.4 times more potent than PGE2 as an antifertility agent in hamsters.{1025}
Catalog No. 50-297-1443 Supplier Cayman Chemical Supplier No. 1481010 MG
17-phenyl trinor PGE2 is a synthetic analog of PGE2. It is an EP1 and EP3 receptor agonist.{3318,1025} 17-phenyl trinor PGE2 is an EP1 and EP3 receptor agonist.1,2 It causes contraction of the guinea pig ileum at a concentration of 11 µM.{3318} It is slightly less potent than PGE2 in stimulating gerbil colon and rat uterus.{1025} With an ED50 value of 350 µg/kg, 17-phenyl trinor PGE2 is 4.4 times more potent than PGE2 as an antifertility agent in hamsters.{1025}