Learn More
Cayman Chemical 17phenyl trinor13 14dihydro P
SDP
Catalog No. 502887510
Encompass_Preferred
17-phenyl trinor-13,14-dihydro PGA2 is a synthetic PG analog whose biological activity has not been widely reported. The PGF2α analog latanoprost, which bears the same lower side chain features, has been approved as a pharmaceutical for the treatment of glaucoma.{5338}
Catalog No. 50-288-7510 Supplier Cayman Chemical Supplier No. 102905 MG
17-phenyl trinor-13,14-dihydro PGA2 is a synthetic PG analog whose biological activity has not been widely reported. The PGF2α analog latanoprost, which bears the same lower side chain features, has been approved as a pharmaceutical for the treatment of glaucoma.{5338}