Learn More
Cayman Chemical 17phenyl TRNor PROSGlndI 10mg
SDP
Catalog No. 502895478
Encompass_Preferred
A form of 17-phenyl trinor PGF modified with a cyclohexane moiety at the alpha carbon; insoluble in aqueous media, soluble in organic solvents; activity at the FP receptor has not been evaluated
Catalog No. 50-289-5478 Supplier Cayman Chemical Supplier No. 900068610 MG
A form of 17-phenyl trinor PGF modified with a cyclohexane moiety at the alpha carbon; insoluble in aqueous media, soluble in organic solvents; activity at the FP receptor has not been evaluated