Learn More
Cayman Chemical 17R 18S-EpETE 25ug
SDP
Catalog No. 502937177
Encompass_Preferred
An EPA metabolite and activator of KCa1.1/BK channels; increases the potassium current amplitude by 15-fold in isolated rat cerebral artery VSMCs at 50 nM; has negative chronotropic effects in NRCMs (EC50 = ~1-2 nM) and prevents calcium-induced increases in NRCM spontaneous beating
Catalog No. 50-293-7177 Supplier Cayman Chemical Supplier No. 2863025 UG
An EPA metabolite and activator of KCa1.1/BK channels; increases the potassium current amplitude by 15-fold in isolated rat cerebral artery VSMCs at 50 nM; has negative chronotropic effects in NRCMs (EC50 = ~1-2 nM) and prevents calcium-induced increases in NRCM spontaneous beating