Learn More
Cayman Chemical 17-trifluoromethylphenyl trinor Prostaglandin F2α, Purity: ≥98%, CAS: 221246-34-0, 10 mg
SDP
Catalog No. 502966095
Encompass_Preferred
A number of 17-phenyl trinor PGF2α derivatives have been approved for the treatment of glaucoma.{8941,9341,8839} Of these, the unsubstituted or meta-substituted aromatic derivatives are the most potent FP receptor agonists.{9668} 17-trifluoromethylphenyl trinor PGF2α bears an aromatic ring which is reminiscent of the trifluoromethyl-phenoxy ring of travoprost ((+)-fluprostenol isopropyl ester). As an ocular hypotensive agent, it would be expected that 17-trifluoromethylphenyl trinor PGF2α would act very much like the free acid of travoprost.
Catalog No. 50-296-6095 Supplier Cayman Chemical Supplier No. 1689010 MG
A number of 17-phenyl trinor PGF2α derivatives have been approved for the treatment of glaucoma.{8941,9341,8839} Of these, the unsubstituted or meta-substituted aromatic derivatives are the most potent FP receptor agonists.{9668} 17-trifluoromethylphenyl trinor PGF2α bears an aromatic ring which is reminiscent of the trifluoromethyl-phenoxy ring of travoprost ((+)-fluprostenol isopropyl ester). As an ocular hypotensive agent, it would be expected that 17-trifluoromethylphenyl trinor PGF2α would act very much like the free acid of travoprost.