Learn More
Cayman Chemical 17-trifluoromethylphenyl-13,14-dihydro trinor Prostaglandin F2α, Purity: ≥98%, CAS: 294856-01-2, 10 mg
SDP
Catalog No. 502967316
Encompass_Preferred
A number of 17-phenyl trinor PGF2α derivatives have been approved for the treatment of glaucoma.{8941,9341,8839} Of these, the ones wherein the 13,14-double bond has been hydrogenated retain relatively good potency, but show a significantly reduced incidence of local irritant side effects.{9736} 17-trifluoromethylphenyl-13,14-dihydro trinor PGF2α bears an aromatic ring which is reminiscent of the trifluoromethyl-phenoxy ring of travoprost ((+)-fluprostenol isopropyl ester). As an ocular hypotensive agent, it would be expected that 17-trifluoromethylphenyl-13,14-dihydro trinor PGF2α would act very much like the free acid of latanoprost.
Catalog No. 50-296-7316 Supplier Cayman Chemical Supplier No. 1689510 MG
A number of 17-phenyl trinor PGF2α derivatives have been approved for the treatment of glaucoma.{8941,9341,8839} Of these, the ones wherein the 13,14-double bond has been hydrogenated retain relatively good potency, but show a significantly reduced incidence of local irritant side effects.{9736} 17-trifluoromethylphenyl-13,14-dihydro trinor PGF2α bears an aromatic ring which is reminiscent of the trifluoromethyl-phenoxy ring of travoprost ((+)-fluprostenol isopropyl ester). As an ocular hypotensive agent, it would be expected that 17-trifluoromethylphenyl-13,14-dihydro trinor PGF2α would act very much like the free acid of latanoprost.