Learn More
Sarstedt Inc 17x110mm Tube, 14mL, Conical Bottom, Polypropylene, 1000/Case
Catalog No. NC9960525 Shop All Sarstedt Inc. Products
Encompass_Preferred
17x110mm Tube 14mL Conical Bottom Polypropylene Graduated No Cap
Catalog No. NC9960525 Supplier Sarstedt Inc Supplier No. 57.527.300
17x110mm Tube 14mL Conical Bottom Polypropylene Graduated No Cap