Learn More
Medchemexpress LLC 18:0 mPEG2000 PE (sodium) | 247925-28-6 | 98.0% | 2808.74 | 10 MG
SDP
Catalog No. 5000438629
Encompass_Preferred
18:0 mPEG2000 PE sodium (DSPE-mPEG2000 sodium) is a conjugate of phospholipid and polyethylene glycol. It functions as a significant PEG lipid component in lipid nanoparticles (LNPs) and is utilized in research concerning gene transfection, drug carriers, and drug delivery. This compound possesses both hydrophilic and hydrophobic properties, enabling it to encapsulate drugs within liposomes to enhance their stability and efficacy.
  • Conjugate of phospholipid and polyethylene glycol
  • Functions as a significant PEG lipid component in lipid nanoparticles (LNPs)
  • Utilized in gene transfection research
  • Used in drug carrier research
  • Used in drug delivery research
  • Possesses both hydrophilic and hydrophobic properties
  • Enables encapsulation of drugs within liposomes to enhance stability and efficacy
  • Intended for research use only
Catalog No. 50-004-38629 Supplier Medchemexpress LLC Supplier No. HY11276010MG
18:0 mPEG2000 PE sodium (DSPE-mPEG2000 sodium) is a conjugate of phospholipid and polyethylene glycol. It functions as a significant PEG lipid component in lipid nanoparticles (LNPs) and is utilized in research concerning gene transfection, drug carriers, and drug delivery. This compound possesses both hydrophilic and hydrophobic properties, enabling it to encapsulate drugs within liposomes to enhance their stability and efficacy.
  • Conjugate of phospholipid and polyethylene glycol
  • Functions as a significant PEG lipid component in lipid nanoparticles (LNPs)
  • Utilized in gene transfection research
  • Used in drug carrier research
  • Used in drug delivery research
  • Possesses both hydrophilic and hydrophobic properties
  • Enables encapsulation of drugs within liposomes to enhance stability and efficacy
  • Intended for research use only