Learn More
Avanti Polar Lipids 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] (ammonium salt), 474922-77-5, MFCD02262268, 25 mg
Catalog No. NC1497487
Encompass
Formula Weight: 2805.497, Purity >99%, Storage Temperature: -20°C. PEG-modified lipids are typically added to phospholipid vesicles used for drug delivery to provide a steric coating on the surface of the membrane to hinder clearing of the particles by the reticuloendothelial system (RES). This prolongs the circulating plasma half-life of the drug. The plasma half-life of the drug can be "tuned" from several hours to days depending on the size of the PEG and fatty acids attached to the lipid anchor.
Catalog No. NC1497487 Supplier Avanti Polar Lipids Supplier No. 880120P25MG
Formula Weight: 2805.497, Purity >99%, Storage Temperature: -20°C. PEG-modified lipids are typically added to phospholipid vesicles used for drug delivery to provide a steric coating on the surface of the membrane to hinder clearing of the particles by the reticuloendothelial system (RES). This prolongs the circulating plasma half-life of the drug. The plasma half-life of the drug can be "tuned" from several hours to days depending on the size of the PEG and fatty acids attached to the lipid anchor.